siRNA Details
Gene Symbol: LINC00534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_051989.1 | 4 | 653 | |||
GenBank mRNA | BG494657.1 | - | 4 | 390 | ||
BG496346.1 | - | 3 | 486 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF132638.1 | - | 469 | Chr. - - [-] on Build |
