siRNA Details
Gene Symbol: LINC00534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_051989.1 | 1 | 143 | |||
GenBank mRNA | BF212685.1 | - | 1 | 145 | ||
BG492574.1 | - | 1 | 94 | |||
BG496346.1 | - | 1 | 102 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_051989.1 | 1 | 143 | |||
GenBank mRNA | BF212685.1 | - | 1 | 145 | ||
BG492574.1 | - | 1 | 94 | |||
BG496346.1 | - | 1 | 102 |