siRNA Details
Gene Symbol: MIR3142HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132748.1 | 1 | 108 | |||
GenBank mRNA | BQ425371.1 | - | 1 | 91 | ||
DQ658414.1 | - | 1 | 108 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MIR3142HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132748.1 | 1 | 108 | |||
GenBank mRNA | BQ425371.1 | - | 1 | 91 | ||
DQ658414.1 | - | 1 | 108 |