siRNA Details
Gene Symbol: CA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121630.1 | 2 | 347 | |||
NR_121631.1 | 2 | 119 | ||||
NR_121632.1 | 2 | 136 | ||||
NR_121633.1 | 2 | 136 | ||||
GenBank mRNA | BM713112.1 | - | 1 | 27 | ||
BM717037.1 | - | 1 | 27 | |||
DA940436.1 | - | 2 | 119 | |||
DB177224.1 | - | 2 | 206 | |||
DB196812.1 | - | 2 | 347 |
