siRNA Details
Gene Symbol: OSGEPL1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102429.1 | 2 | 826 | |||
GenBank mRNA | BX474356.1 | - | 2, 3 | 420 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OSGEPL1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102429.1 | 2 | 826 | |||
GenBank mRNA | BX474356.1 | - | 2, 3 | 420 |