siRNA Details
Gene Symbol: MINCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120682.1 | 1 | 162 | |||
NR_120683.1 | 1 | 162 | ||||
GenBank mRNA | AA992445.1 | - | 1 | 151 | ||
AJ346444.1 | - | 1 | 139 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MINCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120682.1 | 1 | 162 | |||
NR_120683.1 | 1 | 162 | ||||
GenBank mRNA | AA992445.1 | - | 1 | 151 | ||
AJ346444.1 | - | 1 | 139 |