siRNA Details
Gene Symbol: LOC100288181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038925.1 | 3 | 2813 | |||
GenBank mRNA | CR627188.1 | - | 3 | 2814 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100288181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038925.1 | 3 | 2813 | |||
GenBank mRNA | CR627188.1 | - | 3 | 2814 |