siRNA Details
Gene Symbol: LOC105375843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134323.1 | 2 | 357 | |||
GenBank mRNA | AA412543.1 | - | 2 | 358 | ||
CR738863.1 | - | 2 | 356 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105375843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134323.1 | 2 | 357 | |||
GenBank mRNA | AA412543.1 | - | 2 | 358 | ||
CR738863.1 | - | 2 | 356 |