siRNA Details
Gene Symbol: LOC105369486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135100.1 | 1 | 41 | |||
GenBank mRNA | DA016239.1 | - | 1 | 42 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105369486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135100.1 | 1 | 41 | |||
GenBank mRNA | DA016239.1 | - | 1 | 42 |