siRNA Details
Gene Symbol: NAV2-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046672.1 | 1 | 139 | |||
GenBank mRNA | BX112419.1 | - | 1 | 139 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | W44507.1 | - | 139 | Chr. - - [-] on Build |
