siRNA Details
Gene Symbol: KIRREL3-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046986.1 | 2, 3 | 339 | |||
GenBank mRNA | BC031955.1 | - | 2, 3 | 338 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG718299.1 | - | 2, 3 | 343 | Chr. 11 - 126940746 - 126945092 [+] on Build GRCh38 |
