siRNA Details
Gene Symbol: PGGHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011520383.2 | XP_011518685.1 | 2 | 484 | ||
XM_011520384.2 | XP_011518686.1 | 1 | 595 | |||
XM_011520385.2 | XP_011518687.1 | 1 | 598 | |||
XM_011520386.2 | XP_011518688.1 | 1 | 598 | |||
XM_011520387.2 | XP_011518689.1 | 1 | 598 |
