siRNA Details
Gene Symbol: NAV2-AS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_049725.1 | 2 | 2086 | |||
GenBank mRNA | BC040220.1 | - | 2 | 2085 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NAV2-AS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_049725.1 | 2 | 2086 | |||
GenBank mRNA | BC040220.1 | - | 2 | 2085 |