siRNA Details
Gene Symbol: SNHG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003142.2 | 130 | ||||
GenBank mRNA | CR749453.1 | - | 131 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SNHG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003142.2 | 130 | ||||
GenBank mRNA | CR749453.1 | - | 131 |