siRNA Details
Gene Symbol: LOC101928894 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120565.1 | 3 | 502 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM478787.1 | - | 3 | 526 | Chr. 11 - 46171947 - 46177066 [+] on Build GRCh38 |
