siRNA Details
Gene Symbol: LOC100506990 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040091.1 | 1 | 575 | |||
NR_040092.1 | 1 | 575 | ||||
GenBank mRNA | AK095981.1 | - | 1 | 224 | ||
AK129982.1 | - | 1 | 471 | |||
AL539380.3 | - | 1 | 574 | |||
BC037934.1 | - | 1 | 458 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD517001.1 | - | 2 | 110 | Chr. 8 - 12437199 - 12531021 [+] on Build GRCh38 |
