siRNA Details
Gene Symbol: JRKL-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047481.1 | 4 | 391 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL706355.1 | - | 4 | 343 | Chr. 11 - 96447145 - 96506826 [-] on Build GRCh38 |
