siRNA Details
Gene Symbol: USP30-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038996.1 | 1, 2 | 273 | |||
GenBank mRNA | AA143146.1 | - | 1, 2 | 272 | ||
AB371497.1 | - | 1, 2 | 260 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: USP30-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038996.1 | 1, 2 | 273 | |||
GenBank mRNA | AA143146.1 | - | 1, 2 | 272 | ||
AB371497.1 | - | 1, 2 | 260 |