siRNA Details
Gene Symbol: USP30-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038996.1 | 2 | 636 | |||
GenBank mRNA | AB371497.1 | - | 2 | 623 | ||
AI572633.1 | - | 1 | 201 | |||
AW977362.1 | - | 1 | 207 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: USP30-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038996.1 | 2 | 636 | |||
GenBank mRNA | AB371497.1 | - | 2 | 623 | ||
AI572633.1 | - | 1 | 201 | |||
AW977362.1 | - | 1 | 207 |