siRNA Details
Gene Symbol: LOC101928943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120568.1 | 1, 2 | 122 | |||
NR_120569.1 | 1, 2 | 43 | ||||
GenBank mRNA | BX115731.1 | - | 1, 2 | 123 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120568.1 | 1, 2 | 122 | |||
NR_120569.1 | 1, 2 | 43 | ||||
GenBank mRNA | BX115731.1 | - | 1, 2 | 123 |