siRNA Details
Gene Symbol: LOC101928943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120568.1 | 1 | 69 | |||
GenBank mRNA | BX115731.1 | - | 1 | 70 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120568.1 | 1 | 69 | |||
GenBank mRNA | BX115731.1 | - | 1 | 70 |