siRNA Details
Gene Symbol: LOC100130111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135221.1 | 2 | 741 | |||
GenBank mRNA | AK096061.1 | - | 2 | 686 | ||
BC043570.1 | - | 2 | 332 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100130111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135221.1 | 2 | 741 | |||
GenBank mRNA | AK096061.1 | - | 2 | 686 | ||
BC043570.1 | - | 2 | 332 |