siRNA Details
Gene Symbol: LOC101927318 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110589.1 | 1, 2 | 262 | |||
GenBank mRNA | BM931698.1 | - | 1, 2 | 262 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB196366.1 | - | 1, 2 | 261 | Chr. 12 - 49959400 - 49962922 [-] on Build GRCh38 |
