siRNA Details
Gene Symbol: PSMA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029434.1 | 2, 3 | 261 | |||
GenBank mRNA | BC034618.1 | - | 2, 3 | 253 | ||
BC104151.1 | - | 3, 4 | 487 | |||
BC104152.1 | - | 2, 3 | 256 | |||
BC112973.1 | - | 2, 3 | 264 |
