siRNA Details
Gene Symbol: PSMA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029434.1 | 4 | 443 | |||
NR_029435.1 | 3 | 402 | ||||
GenBank mRNA | AK055868.1 | - | 4 | 2306 | ||
BC034618.1 | - | 4 | 435 | |||
BU533562.1 | - | 4 | 280 | |||
BX460997.2 | - | 3 | 386 | |||
BX482479.1 | - | 3 | 377 | |||
CR627086.1 | - | 3 | 394 |
