siRNA Details
Gene Symbol: LOC100506125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110019.1 | 2 | 166 | |||
NR_110020.1 | 2 | 200 | ||||
GenBank mRNA | BX098326.1 | - | 2 | 166 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100506125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110019.1 | 2 | 166 | |||
NR_110020.1 | 2 | 200 | ||||
GenBank mRNA | BX098326.1 | - | 2 | 166 |