siRNA Details
Gene Symbol: LOC100506844 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038269.1 | 3 | 535 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100506844 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038269.1 | 3 | 535 |