siRNA Details
Gene Symbol: LOC101927062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110033.1 | 4 | 485 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX101084.1 | - | 4 | 472 | Chr. 14 - 26775495 - 26822107 [-] on Build GRCh38 |
