siRNA Details
Gene Symbol: BCDIN3D-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027499.1 | 3 | 2148 | |||
NR_027500.1 | 4 | 658 | ||||
NR_027501.1 | 3 | 2144 | ||||
GenBank mRNA | AK055142.1 | - | 3 | 2148 | ||
BC014925.1 | - | 3 | 2079 |
