siRNA Details
Gene Symbol: RHOXF1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131238.1 | 4, 5 | 888 | |||
GenBank mRNA | AK123976.1 | - | 4, 5 | 887 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RHOXF1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131238.1 | 4, 5 | 888 | |||
GenBank mRNA | AK123976.1 | - | 4, 5 | 887 |