siRNA Details
Gene Symbol: LINC00645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039992.2 | 4, 5 | 327 | |||
GenBank mRNA | BC042069.1 | - | 1, 2 | 95 | ||
BX538073.1 | - | 4, 5 | 304 | |||
BX538140.1 | - | 3, 4 | 276 |
