siRNA Details
Gene Symbol: DIO2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038355.1 | 8 | 1232 | |||
GenBank mRNA | BC039670.1 | - | 8 | 1232 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DIO2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038355.1 | 8 | 1232 | |||
GenBank mRNA | BC039670.1 | - | 8 | 1232 |