siRNA Details
Gene Symbol: LRP1-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131938.1 | 1 | 52 | |||
NR_131939.1 | 1 | 52 | ||||
GenBank mRNA | BM714117.1 | - | 1 | 51 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LRP1-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131938.1 | 1 | 52 | |||
NR_131939.1 | 1 | 52 | ||||
GenBank mRNA | BM714117.1 | - | 1 | 51 |