siRNA Details
Gene Symbol: LINC00648 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039996.1 | 1, 2 | 52 | |||
GenBank mRNA | AK056212.1 | - | 1, 2 | 52 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00648 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039996.1 | 1, 2 | 52 | |||
GenBank mRNA | AK056212.1 | - | 1, 2 | 52 |