siRNA Details
Gene Symbol: LINC00929 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038851.1 | 1 | 180 | |||
NR_038852.1 | 1 | 180 | ||||
GenBank mRNA | BC038777.1 | - | 1 | 168 | ||
BX117286.1 | - | 1 | 180 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00929 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038851.1 | 1 | 180 | |||
NR_038852.1 | 1 | 180 | ||||
GenBank mRNA | BC038777.1 | - | 1 | 168 | ||
BX117286.1 | - | 1 | 180 |