siRNA Details
Gene Symbol: LOC101928909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110134.1 | 2 | 331 | |||
GenBank mRNA | BC028598.1 | - | 2 | 325 | ||
BC028746.1 | - | 2 | 325 | |||
HY025890.1 | - | 2 | 332 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110134.1 | 2 | 331 | |||
GenBank mRNA | BC028598.1 | - | 2 | 325 | ||
BC028746.1 | - | 2 | 325 | |||
HY025890.1 | - | 2 | 332 |