siRNA Details
Gene Symbol: LOC101928909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110134.1 | 5 | 1077 | |||
GenBank mRNA | AA639311.1 | - | 1 | 372 | ||
BC028598.1 | - | 5 | 1071 | |||
BC028746.1 | - | 5 | 1071 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110134.1 | 5 | 1077 | |||
GenBank mRNA | AA639311.1 | - | 1 | 372 | ||
BC028598.1 | - | 5 | 1071 | |||
BC028746.1 | - | 5 | 1071 |