siRNA Details
Gene Symbol: LINC00927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033833.1 | 3 | 2691 | |||
GenBank mRNA | BC040586.1 | - | 3 | 2654 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033833.1 | 3 | 2691 | |||
GenBank mRNA | BC040586.1 | - | 3 | 2654 |