siRNA Details
Gene Symbol: LOC101927286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120324.1 | 3 | 450 | |||
GenBank mRNA | BC038379.1 | - | 3 | 450 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI756744.1 | - | 3 | 449 | Chr. 15 - 97371289 - 97521811 [-] on Build GRCh38 |
