siRNA Details
Gene Symbol: IQCH-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040051.1 | 6 | 1277 | |||
NR_040052.1 | 6 | 1073 | ||||
GenBank mRNA | BC047537.1 | - | 6 | 1073 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX451373.1 | - | 2 | 255 | Chr. 15 - 67407629 - 67421391 [-] on Build GRCh38 |
