siRNA Details
Gene Symbol: IQCH-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040051.1 | 4 | 777 | |||
GenBank mRNA | BX332831.2 | - | 4 | 570 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IQCH-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040051.1 | 4 | 777 | |||
GenBank mRNA | BX332831.2 | - | 4 | 570 |