siRNA Details
Gene Symbol: IQCH-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040051.1 | 3 | 584 | |||
NR_040052.1 | 3 | 584 | ||||
GenBank mRNA | BC047537.1 | - | 3 | 584 | ||
BX332831.2 | - | 3 | 377 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IQCH-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040051.1 | 3 | 584 | |||
NR_040052.1 | 3 | 584 | ||||
GenBank mRNA | BC047537.1 | - | 3 | 584 | ||
BX332831.2 | - | 3 | 377 |