siRNA Details
Gene Symbol: IL21R-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037158.1 | 2, 3 | 478 | |||
GenBank mRNA | AK093371.1 | - | 2, 3 | 478 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB033419.1 | - | 2, 3 | 478 | Chr. 16 - 27449746 - 27453393 [-] on Build GRCh38 |
