siRNA Details
Gene Symbol: LOC101929224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110787.1 | 3 | 635 | |||
GenBank mRNA | BC042016.1 | - | 3 | 635 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD240151.1 | - | 472 | Chr. - - [-] on Build |
