siRNA Details
Gene Symbol: SNHG19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132114.1 | 2 | 253 | |||
GenBank mRNA | AA535774.1 | - | 2 | 248 | ||
BM856089.1 | - | 2 | 253 | |||
DA194369.1 | - | 1 | 473 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA095544.1 | - | 1 | 473 | Chr. 16 - 2154799 - 2155358 [-] on Build GRCh38 |
