siRNA Details
Gene Symbol: LINC01043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135321.1 | 5895 | ||||
GenBank mRNA | BX649107.1 | - | 5895 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DR763502.1 | - | 1 | 366 | Chr. 13 - 112318996 - 112321193 [+] on Build GRCh38 |
