siRNA Details
Gene Symbol: LOC101928167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110800.1 | 2 | 676 | |||
GenBank mRNA | BC034434.1 | - | 2 | 677 | ||
BC036634.2 | - | 2 | 666 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110800.1 | 2 | 676 | |||
GenBank mRNA | BC034434.1 | - | 2 | 677 | ||
BC036634.2 | - | 2 | 666 |