siRNA Details
Gene Symbol: ATP2A1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046287.1 | 2 | 448 | |||
NR_046288.1 | 2 | 445 | ||||
NR_046289.1 | 2 | 395 | ||||
NR_046290.1 | 2 | 392 | ||||
GenBank mRNA | BX099641.1 | - | 2 | 422 | ||
CK823012.1 | - | 2 | 394 |
