siRNA Details
Gene Symbol: PWRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026647.1 | 1 | 4333 | |||
GenBank mRNA | BC032951.1 | - | 2 | 3459 | ||
BC070120.1 | - | 2 | 3454 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PWRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026647.1 | 1 | 4333 | |||
GenBank mRNA | BC032951.1 | - | 2 | 3459 | ||
BC070120.1 | - | 2 | 3454 |