siRNA Details
Gene Symbol: ERVK13-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040023.1 | 3 | 3439 | |||
GenBank mRNA | AK091794.1 | - | 3 | 1231 | ||
AK095440.1 | - | 1 | 1036 | |||
AK097014.1 | - | 2 | 971 | |||
AK131509.1 | - | 1 | 2065 | |||
AK292678.1 | - | 1 | 12 | |||
AX747190.1 | - | 3 | 1231 | |||
BC043252.1 | - | 3 | 3441 |
